Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319214.1 | NP_001306143.1 | 8 | 1150 | ||
NM_005040.3 | NP_005031.1 | 9 | 1512 | |||
NM_199418.3 | NP_955450.2 | 10 | 1575 | |||
XM_005274093.1 | XP_005274150.1 | 9 | 1242 | |||
GenBank mRNA | AB451270.1 | - | 9 | 1311 | ||
AB451397.1 | - | 9 | 1311 | |||
AK090847.1 | - | 3 | 1808 | |||
AK091786.1 | - | 10 | 1402 | |||
AK302386.1 | - | 8 | 1101 | |||
AK312919.1 | - | 9 | 1333 | |||
BC001500.2 | - | 9 | 1330 | 3510081 | ||
BX537480.1 | - | 1 | 6331 | |||
L13977.1 | - | 9 | 1340 |