Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015596.1 | NP_056411.1 | 5 | 873 | ||
GenBank mRNA | AY923171.1 | - | 5 | 662 | ||
AY923172.1 | - | 6 | 1229 | |||
AY923173.1 | - | 6 | 938 | |||
AY923174.1 | - | 5 | 1145 | |||
AY923175.1 | - | 5 | 854 | |||
BC069334.1 | - | 5 | 874 | 7262205 | ||
BC069543.1 | - | 5 | 873 | 7262217 |