Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRPM6 SIRNA Assays ›
          Gene Symbol:
          TRPM6
          Gene Name:
          transient receptor potential cation channel subfamily M member 6
          Gene Aliases:
          CHAK2, HMGX, HOMG, HOMG1, HSH
          Chromosome Location:
          Chr.9: 74722495 - 74888094 on Build GRCh38
          UniGene ID:
          Hs.272225
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104671
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRPM6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001177310.1 NP_001170781.1 10, 11 1271
          NM_001177311.1 NP_001170782.1 10, 11 1298
          NM_017662.4 NP_060132.3 10, 11 1452
          XM_011518244.1 XP_011516546.1 10, 11 1452
          XM_011518245.1 XP_011516547.1 10, 11 1452
          XM_011518246.1 XP_011516548.1 10, 11 1452
          XM_011518247.1 XP_011516549.1 10, 11 1452
          XM_011518248.1 XP_011516550.1 10, 11 1452
          XM_011518249.1 XP_011516551.1 10, 11 1452
          XM_011518250.1 XP_011516552.1 10, 11 1452
          XM_011518251.2 XP_011516553.1 5, 6 703
          XM_011518252.2 XP_011516554.1 10, 11 1452
          XM_011518255.2 XP_011516557.1 10, 11 1452
          XM_017014287.1 XP_016869776.1 10, 11 1452
          XM_017014288.1 XP_016869777.1 10, 11 1452
          XM_017014289.1 XP_016869778.1 10, 11 1452
          GenBank mRNA AF350881.2 - 10, 11 1452
          AF448232.1 - 10, 11 1214
          AK026281.1 - 4, 5 519
          AY333282.1 - 10, 11 1277
          AY333283.1 - 10, 11 1271
          AY333284.1 - 10, 11 1298
          AY333285.1 - 10, 11 1277
          AY333286.1 - 10, 11 1277
          AY333287.1 - 10, 11 1277

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00214306_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          response to toxic substance
          protein tetramerization
          calcium ion transmembrane transport
          protein serine/threonine kinase activity
          calcium channel activity
          protein binding
          ATP binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6f7b98477d-hxtzg:80/100.66.130.62:80.
          git-commit: 5163bf559e7ad0ebe9326a88094353fc6888890d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline