Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRPM3 SIRNA Assays ›
          Gene Symbol:
          TRPM3
          Gene Name:
          transient receptor potential cation channel subfamily M member 3
          Gene Aliases:
          GON-2, LTRPC3, MLSN2
          Chromosome Location:
          Chr.9: 70529063 - 71446989 on Build GRCh38
          UniGene ID:
          Hs.47288
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 104752

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRPM3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001007470.1 NP_001007471.1 5 687
          NM_001007471.2 NP_001007472.2 5 1035
          NM_020952.4 NP_066003.3 5 687
          NM_024971.5 NP_079247.5 5 687
          NM_206944.3 NP_996827.3 5 687
          NM_206945.3 NP_996828.3 5 687
          NM_206946.3 NP_996829.3 5 687
          NM_206947.3 NP_996830.3 5 687
          NM_206948.2 NP_996831.1 5 687
          XM_011519035.2 XP_011517337.1 5 912
          XM_011519036.2 XP_011517338.1 5 3837
          XM_011519037.2 XP_011517339.1 5 3823
          XM_011519038.2 XP_011517340.1 5 3820
          XM_011519039.2 XP_011517341.1 5 3821
          XM_011519040.2 XP_011517342.1 5 918
          XM_011519041.2 XP_011517343.1 5 3820
          XM_011519042.2 XP_011517344.1 5 918
          XM_011519043.2 XP_011517345.1 5 951
          XM_011519044.2 XP_011517346.1 5 3821
          XM_011519045.2 XP_011517347.1 5 912
          XM_011519046.2 XP_011517348.1 5 2615
          XM_011519047.2 XP_011517349.1 5 2585
          XM_017015141.1 XP_016870630.1 5 912
          XM_017015142.1 XP_016870631.1 5 912
          XM_017015143.1 XP_016870632.1 5 912
          XM_017015144.1 XP_016870633.1 5 678
          XM_017015145.1 XP_016870634.1 5 1356
          XM_017015146.1 XP_016870635.1 5 754
          XM_017015147.1 XP_016870636.1 5 2789
          XM_017015148.1 XP_016870637.1 5 912
          XM_017015149.1 XP_016870638.1 5 912
          XM_017015150.1 XP_016870639.1 5 912
          XM_017015151.1 XP_016870640.1 5 930
          XM_017015152.1 XP_016870641.1 5 2590
          XM_017015153.1 XP_016870642.1 5 922
          XM_017015154.1 XP_016870643.1 5 1058
          XM_017015155.1 XP_016870644.1 5 2587
          XM_017015156.1 XP_016870645.1 1 110
          XM_017015157.1 XP_016870646.1 5 2585
          XM_017015158.1 XP_016870647.1 5 2583
          XM_017015159.1 XP_016870648.1 5 678
          XM_017015160.1 XP_016870649.1 5 768
          XM_017015161.1 XP_016870650.1 5 912
          GenBank mRNA AB099661.1 - 2 332
          AB099662.1 - 2 332
          AB099663.1 - 2 332
          AB099664.1 - 2 332
          AB099665.1 - 2 332
          AF325212.1 - 5 658
          AF536748.1 - 2 332
          AF536749.1 - 2 332
          AF536750.1 - 2 332
          AF536751.1 - 2 332
          AF536752.1 - 2 332
          AF536753.1 - 2 332
          AJ505025.1 - 5 1035
          AJ505026.1 - 5 1035
          AK021788.1 - 5 678
          AK225732.1 - 5 678
          AK308682.1 - 5 863
          AL136545.1 - 5 868
          BC022454.2 - 4 617
          BC067733.1 - 5 649
          BC094699.1 - 5 681
          BC121821.1 - 3 533
          BC134414.1 - 3 536
          BC142972.1 - 5 704
          KF987075.1 - 5 743

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cation transport
          detection of temperature stimulus
          sensory perception of temperature stimulus
          protein tetramerization
          calcium ion transmembrane transport
          calcium activated cation channel activity
          cation channel activity
          calcium channel activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix