Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007271.1 | NP_001007272.1 | 8 | 1736 | ||
NM_001007272.1 | NP_001007273.1 | 4 | 1011 | |||
NM_001007273.1 | NP_001007274.1 | 6 | 1310 | |||
NM_001320842.1 | NP_001307771.1 | 7 | 1528 | |||
NM_001320843.1 | NP_001307772.1 | 5 | 1106 | |||
NM_016364.3 | NP_057448.3 | 4 | 869 | |||
XM_005269883.3 | XP_005269940.1 | 7 | 1506 | |||
XM_005269884.4 | XP_005269941.3 | 4 | 1298 | |||
XM_005269887.1 | XP_005269944.1 | 7 | 1608 | |||
XM_005269890.1 | XP_005269947.1 | 5 | 1208 | |||
XM_011539853.1 | XP_011538155.1 | 8 | 1745 | |||
XM_011539854.2 | XP_011538156.1 | 6 | 1284 | |||
XM_011539855.1 | XP_011538157.1 | 4 | 987 | |||
XM_011539856.2 | XP_011538158.1 | 6 | 1288 | |||
XM_017016313.1 | XP_016871802.1 | 4 | 961 | |||
GenBank mRNA | AB027004.1 | - | 4 | 883 | ||
AB103375.1 | - | 4 | 883 | |||
AK057012.1 | - | 5 | 1079 | |||
AK127886.1 | - | 7 | 1528 | |||
AY040091.1 | - | 8 | 1700 | |||
AY674051.1 | - | 8 | 1709 | |||
BC009778.1 | - | 5 | 995 | 4047963 |