Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL12RB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258215.1 | NP_001245144.1 | 12, 13 | 2337 | ||
NM_001559.2 | NP_001550.1 | 14, 15 | 2595 | |||
XM_005270825.2 | XP_005270882.1 | 15, 16 | 2696 | |||
XM_005270827.2 | XP_005270884.1 | 15, 16 | 2692 | |||
XM_005270828.3 | XP_005270885.1 | 14, 15 | 2604 | |||
XM_006710617.2 | XP_006710680.1 | 13, 14 | 2438 | |||
XM_011541383.2 | XP_011539685.1 | 15, 16 | 2148 | |||
XM_011541384.2 | XP_011539686.1 | 14, 15 | 2537 | |||
XM_017001202.1 | XP_016856691.1 | 15, 16 | 2696 | |||
GenBank mRNA | AK300020.1 | - | 3, 4 | 394 | ||
BC104772.1 | - | 13, 14 | 2005 | 8143775 | ||
BC104774.1 | - | 13, 14 | 2005 | 8143777 | ||
BC143249.1 | - | 11, 12 | 1747 | 9051751 | ||
U64198.1 | - | 14, 15 | 2595 |