Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF266 SIRNA Assays ›
Gene Symbol:
ZNF266
Gene Name:
zinc finger protein 266
Gene Aliases:
HZF1
Chromosome Location:
Chr.19: 9412426 - 9435578 on Build GRCh38
UniGene ID:
Hs.656185
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
108074
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF266
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271314.1
NP_001258243.1
11
2000
NM_006631.3
NP_006622.2
11
2051
XM_005259716.3
XP_005259773.1
8
1717
XM_006722620.2
XP_006722683.1
11
1783
XM_006722621.2
XP_006722684.1
10
1689
XM_006722622.2
XP_006722685.1
9
1528
XM_006722623.3
XP_006722686.1
8
1463
XM_006722627.2
XP_006722690.1
10
2380
XM_006722630.2
XP_006722693.1
9
1694
XM_011527644.1
XP_011525946.1
10
1699
XM_011527647.1
XP_011525949.1
11
2238
XM_011527648.1
XP_011525950.1
11
2234
XM_011527650.1
XP_011525952.1
11
2037
XM_017026154.1
XP_016881643.1
11
1808
XM_017026155.1
XP_016881644.1
11
1805
XM_017026156.1
XP_016881645.1
12
2108
XM_017026157.1
XP_016881646.1
10
2383
XM_017026158.1
XP_016881647.1
12
2079
XM_017026159.1
XP_016881648.1
12
2101
XM_017026160.1
XP_016881649.1
9
2274
XM_017026161.1
XP_016881650.1
9
1979
XM_017026162.1
XP_016881651.1
10
1855
XM_017026163.1
XP_016881652.1
10
1824
XM_017026164.1
XP_016881653.1
8
1629
XM_017026165.1
XP_016881654.1
11
1798
XM_017026166.1
XP_016881655.1
8
1619
XM_017026167.1
XP_016881656.1
8
1453
XM_017026168.1
XP_016881657.1
9
2279
XM_017026169.1
XP_016881658.1
7
1825
XM_017026170.1
XP_016881659.1
8
1439
XM_017026171.1
XP_016881660.1
7
1374
XM_017026172.1
XP_016881661.1
7
1364
XM_017026173.1
XP_016881662.1
10
1719
GenBank mRNA
AB095928.1
-
5
2836
AK027480.1
-
1
288
AL833471.1
-
4
956
AL833503.1
-
11
1730
AY376240.1
-
10
1756
BC071628.1
-
1
307
BC112919.1
-
9
1940
7502631
BC126255.1
-
4
1026
8991971
BC143940.1
-
4
1026
BC146837.1
-
5
896
9056764
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix