Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM23 SIRNA Assays ›
Gene Symbol:
RBM23
Gene Name:
RNA binding motif protein 23
Gene Aliases:
CAPERbeta, PP239, RNPC4
Chromosome Location:
Chr.14: 22900645 - 22919187 on Build GRCh38
UniGene ID:
Hs.4997
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
109050
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM23
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077351.1
NP_001070819.1
8, 9
933
NM_001077352.1
NP_001070820.1
6, 7
831
NM_001308044.1
NP_001294973.1
6, 7
772
NM_018107.4
NP_060577.3
7, 8
885
XM_011536890.2
XP_011535192.1
9, 10
1016
XM_011536892.2
XP_011535194.1
8, 9
967
XM_011536893.2
XP_011535195.1
8, 9
968
XM_011536894.2
XP_011535196.1
8, 9
962
XM_011536895.2
XP_011535197.1
7, 8
938
XM_011536896.2
XP_011535198.1
7, 8
914
XM_011536897.2
XP_011535199.1
7, 8
1300
XM_011536900.1
XP_011535202.1
7, 8
876
XM_011536902.1
XP_011535204.1
7, 8
782
XM_011536903.1
XP_011535205.1
5, 6
658
XM_011536904.1
XP_011535206.1
6, 7
715
XM_011536905.1
XP_011535207.1
5, 6
713
XM_011536906.1
XP_011535208.1
4, 5
604
XM_017021398.1
XP_016876887.1
8, 9
970
XM_017021399.1
XP_016876888.1
7, 8
917
XM_017021400.1
XP_016876889.1
7, 8
835
XM_017021401.1
XP_016876890.1
8, 9
930
XM_017021402.1
XP_016876891.1
8, 9
841
XM_017021403.1
XP_016876892.1
7, 8
859
XM_017021404.1
XP_016876893.1
7, 8
809
XM_017021405.1
XP_016876894.1
7, 8
876
XM_017021406.1
XP_016876895.1
8, 9
1348
XM_017021407.1
XP_016876896.1
8, 9
857
XM_017021408.1
XP_016876897.1
6, 7
793
XM_017021409.1
XP_016876898.1
7, 8
1294
XM_017021410.1
XP_016876899.1
6, 7
815
XM_017021411.1
XP_016876900.1
6, 7
1246
XM_017021412.1
XP_016876901.1
8, 9
968
XM_017021413.1
XP_016876902.1
7, 8
744
XM_017021414.1
XP_016876903.1
7, 8
782
XM_017021415.1
XP_016876904.1
6, 7
763
XM_017021416.1
XP_016876905.1
5, 6
658
XM_017021417.1
XP_016876906.1
6, 7
715
XM_017021418.1
XP_016876907.1
5, 6
713
XM_017021419.1
XP_016876908.1
4, 5
604
XM_017021420.1
XP_016876909.1
5, 6
1133
GenBank mRNA
AF087905.1
-
7, 8
836
AF275678.1
-
8, 9
888
AK001344.1
-
6, 7
781
AK125593.1
-
8, 9
895
AK225792.1
-
8, 9
895
AK303668.1
-
6, 7
734
AK308055.1
-
7, 8
848
AK315620.1
-
7, 8
847
AL834198.1
-
8, 9
894
BC002566.2
-
7, 8
863
3138909
BC024208.2
-
7, 8
822
2960467
BC106012.1
-
5, 6
607
BX161440.1
-
8, 9
866
CR457257.1
-
6, 7
686
KU178687.1
-
6, 7
686
KU178688.1
-
7, 8
734
KU178689.1
-
5, 6
632
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mRNA processing
nucleotide binding
RNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0