Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPALPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001316951.1 | NP_001303880.1 | 3, 4 | 456 | ||
NM_001316952.1 | NP_001303881.1 | 3, 4 | 456 | |||
NM_018559.3 | NP_061029.2 | 3, 4 | 456 | |||
XM_005266444.3 | XP_005266501.1 | 3, 4 | 448 | |||
XM_011535144.2 | XP_011533446.1 | 3, 4 | 394 | |||
GenBank mRNA | AB051491.1 | - | 3, 4 | 366 | ||
AF145122.1 | - | 3, 4 | 390 | |||
AF245045.1 | - | 2, 3 | 242 | |||
AK292493.1 | - | 3, 4 | 429 | |||
AK304669.1 | - | 3, 4 | 446 | |||
BC017776.1 | - | 3, 4 | 441 | |||
BC032333.1 | - | 3, 4 | 382 | 5169953 | ||
BC039586.1 | - | 3, 4 | 434 | 5722236 | ||
DC333111.1 | - | 3, 4 | 456 |