Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289112.1 | NP_001276041.1 | 12 | 1354 | ||
NM_130808.2 | NP_570720.1 | 12 | 1982 | |||
NM_153429.1 | NP_702907.1 | 13 | 1342 | |||
XM_011512406.2 | XP_011510708.1 | 11 | 1129 | |||
XM_011512407.2 | XP_011510709.1 | 11 | 1386 | |||
XM_011512408.2 | XP_011510710.1 | 11 | 2514 | |||
XM_017005693.1 | XP_016861182.1 | 15 | 2735 | |||
XM_017005694.1 | XP_016861183.1 | 11 | 1106 | |||
XM_017005695.1 | XP_016861184.1 | 14 | 1605 | |||
XM_017005696.1 | XP_016861185.1 | 6 | 499 | |||
GenBank mRNA | AF465770.1 | - | 15 | 1682 | ||
AF465771.1 | - | 13 | 1342 | |||
AK056175.1 | - | 12 | 1454 | |||
AK128117.1 | - | 16 | 3232 | |||
AK295557.1 | - | 12 | 1269 | |||
BC014396.1 | - | 12 | 1540 | 3162811 | ||
BC075046.2 | - | 12 | 1131 | 30915576 |