Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PPFIA2 SIRNA Assays ›
          Gene Symbol:
          PPFIA2
          Gene Name:
          PTPRF interacting protein alpha 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 81257975 - 81759748 on Build GRCh38
          UniGene ID:
          Hs.506216
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 111323

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PPFIA2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001220473.2 NP_001207402.1 2 405
          NM_001220474.2 NP_001207403.1 2 405
          NM_001220475.2 NP_001207404.1 2 217
          NM_001220476.2 NP_001207405.1 2 217
          NM_003625.4 NP_003616.2 3 268
          XM_011538906.2 XP_011537208.1 2 399
          XM_011538907.2 XP_011537209.1 2 399
          XM_017020079.1 XP_016875568.1 3 252
          XM_017020080.1 XP_016875569.1 2 399
          XM_017020081.1 XP_016875570.1 2 399
          XM_017020082.1 XP_016875571.1 3 252
          XM_017020083.1 XP_016875572.1 2 399
          XM_017020084.1 XP_016875573.1 2 399
          XM_017020085.1 XP_016875574.1 2 399
          XM_017020086.1 XP_016875575.1 3 252
          XM_017020087.1 XP_016875576.1 2 386
          XM_017020088.1 XP_016875577.1 3 686
          XM_017020089.1 XP_016875578.1 2 399
          XM_017020090.1 XP_016875579.1 3 252
          XM_017020091.1 XP_016875580.1 2 399
          XM_017020092.1 XP_016875581.1 2 399
          XM_017020093.1 XP_016875582.1 2 399
          XM_017020094.1 XP_016875583.1 3 252
          XM_017020095.1 XP_016875584.1 2 399
          XM_017020096.1 XP_016875585.1 2 399
          XM_017020097.1 XP_016875586.1 3 252
          XM_017020098.1 XP_016875587.1 3 252
          XM_017020099.1 XP_016875588.1 2 399
          XM_017020100.1 XP_016875589.1 2 399
          XM_017020101.1 XP_016875590.1 3 252
          XM_017020102.1 XP_016875591.1 2 386
          XM_017020103.1 XP_016875592.1 2 399
          XM_017020104.1 XP_016875593.1 2 399
          XM_017020105.1 XP_016875594.1 2 399
          XM_017020106.1 XP_016875595.1 2 399
          XM_017020107.1 XP_016875596.1 3 252
          XM_017020108.1 XP_016875597.1 3 252
          XM_017020109.1 XP_016875598.1 2 399
          XM_017020110.1 XP_016875599.1 3 686
          XM_017020111.1 XP_016875600.1 2 399
          GenBank mRNA AB210009.1 - 2 403
          AF034799.1 - 3 276
          AK126971.1 - 2 365
          AK307524.1 - 3 252
          BC104912.1 - 2 201 8143915
          BC143485.1 - 2 201 9051993
          DA499362.1 - 2 406

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          neurotransmitter secretion
          glutamate secretion
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline