Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZDHHC3 SIRNA Assays ›
          Gene Symbol:
          ZDHHC3
          Gene Name:
          zinc finger DHHC-type containing 3
          Gene Aliases:
          DHHC-3, GODZ, ZNF373
          Chromosome Location:
          Chr.3: 44915261 - 44976190 on Build GRCh38
          UniGene ID:
          Hs.61430
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 113536

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZDHHC3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001135179.1 NP_001128651.1 2 519
          NM_016598.2 NP_057682.1 2 519
          XM_005265209.1 XP_005265266.1 2 519
          XM_005265212.2 XP_005265269.1 2 527
          XM_006713189.1 XP_006713252.1 2 519
          XM_006713192.2 XP_006713255.1 2 519
          XM_006713193.2 XP_006713256.1 2 520
          XM_006713194.3 XP_006713257.1 2 498
          XM_011533797.1 XP_011532099.1 2 519
          XM_011533800.2 XP_011532102.1 2 516
          XM_011533801.2 XP_011532103.1 2 519
          XM_017006557.1 XP_016862046.1 2 519
          XM_017006558.1 XP_016862047.1 3 565
          XM_017006559.1 XP_016862048.1 2 614
          XM_017006560.1 XP_016862049.1 2 519
          XM_017006561.1 XP_016862050.1 2 519
          XM_017006562.1 XP_016862051.1 2 519
          XM_017006563.1 XP_016862052.1 2 519
          XM_017006564.1 XP_016862053.1 2 519
          XM_017006565.1 XP_016862054.1 2 504
          XM_017006566.1 XP_016862055.1 3 557
          XM_017006567.1 XP_016862056.1 3 595
          XM_017006568.1 XP_016862057.1 1 24
          XM_017006569.1 XP_016862058.1 2 496
          XM_017006570.1 XP_016862059.1 2 502
          XM_017006571.1 XP_016862060.1 2 511
          GenBank mRNA AF247703.1 - 2 459
          AF441791.1 - 1 245
          BC015467.1 - 2 446 3852521
          DB636483.1 - 2 519

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001039014, XM_008766715, XM_008766714, XM_006244188, XM_008766716, and others.
          This siRNA also targets the following transcripts in Mouse: NM_026917, BC052464, AB088044, AK018755, AK169197, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting
          protein palmitoylation
          zinc ion binding
          palmitoyltransferase activity
          protein-cysteine S-palmitoyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0