Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPRD SIRNA Assays ›
          Gene Symbol:
          PTPRD
          Gene Name:
          protein tyrosine phosphatase, receptor type D
          Gene Aliases:
          HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
          Chromosome Location:
          Chr.9: 8314246 - 10612840 on Build GRCh38
          UniGene ID:
          Hs.446083
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 114290

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPRD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001040712.2 NP_001035802.1 1, 2 178
          NM_001171025.1 NP_001164496.1 1, 2 178
          NM_002839.3 NP_002830.1 12, 13 818
          NM_130391.3 NP_569075.2 1, 2 178
          NM_130392.3 NP_569076.2 1, 2 178
          NM_130393.3 NP_569077.2 1, 2 178
          XM_006716817.3 XP_006716880.1 11, 12 1116
          XM_006716823.2 XP_006716886.1 11, 12 1110
          XM_006716825.3 XP_006716888.1 11, 12 1112
          XM_006716827.3 XP_006716890.1 11, 12 1109
          XM_006716832.3 XP_006716895.1 11, 12 1104
          XM_006716833.3 XP_006716896.1 11, 12 1106
          XM_006716834.3 XP_006716897.1 11, 12 1107
          XM_006716835.3 XP_006716898.1 11, 12 1106
          XM_006716837.3 XP_006716900.1 11, 12 1104
          XM_006716838.3 XP_006716901.1 11, 12 1106
          XM_006716839.3 XP_006716902.1 11, 12 1104
          XM_011517992.2 XP_011516294.1 11, 12 1109
          XM_017014958.1 XP_016870447.1 11, 12 1115
          XM_017014959.1 XP_016870448.1 12, 13 1187
          XM_017014960.1 XP_016870449.1 10, 11 1076
          XM_017014961.1 XP_016870450.1 10, 11 1060
          XM_017014962.1 XP_016870451.1 2, 3 248
          XM_017014963.1 XP_016870452.1 11, 12 1115
          XM_017014964.1 XP_016870453.1 11, 12 1114
          XM_017014965.1 XP_016870454.1 11, 12 1117
          XM_017014966.1 XP_016870455.1 11, 12 1115
          XM_017014967.1 XP_016870456.1 11, 12 1113
          XM_017014968.1 XP_016870457.1 11, 12 1112
          XM_017014969.1 XP_016870458.1 11, 12 1115
          XM_017014970.1 XP_016870459.1 11, 12 1115
          XM_017014971.1 XP_016870460.1 11, 12 1113
          XM_017014972.1 XP_016870461.1 11, 12 1115
          XM_017014973.1 XP_016870462.1 11, 12 1112
          XM_017014974.1 XP_016870463.1 11, 12 1115
          XM_017014975.1 XP_016870464.1 11, 12 1110
          XM_017014976.1 XP_016870465.1 11, 12 1112
          XM_017014977.1 XP_016870466.1 11, 12 1113
          XM_017014978.1 XP_016870467.1 11, 12 1112
          XM_017014979.1 XP_016870468.1 11, 12 1114
          XM_017014980.1 XP_016870469.1 11, 12 1109
          XM_017014981.1 XP_016870470.1 11, 12 1104
          XM_017014982.1 XP_016870471.1 11, 12 1111
          XM_017014983.1 XP_016870472.1 11, 12 1111
          XM_017014984.1 XP_016870473.1 11, 12 1111
          XM_017014985.1 XP_016870474.1 11, 12 1108
          XM_017014986.1 XP_016870475.1 11, 12 1107
          XM_017014987.1 XP_016870476.1 11, 12 1107
          XM_017014988.1 XP_016870477.1 11, 12 1107
          XM_017014989.1 XP_016870478.1 11, 12 1107
          XM_017014990.1 XP_016870479.1 11, 12 1106
          XM_017014991.1 XP_016870480.1 11, 12 1106
          XM_017014992.1 XP_016870481.1 11, 12 1106
          XM_017014993.1 XP_016870482.1 11, 12 1104
          XM_017014994.1 XP_016870483.1 11, 12 1104
          XM_017014995.1 XP_016870484.1 11, 12 1110
          GenBank mRNA AB211400.1 - 11, 12 786
          BC028038.1 - 890
          BC106713.1 - 1, 2 126 40027581
          BC106714.1 - 1, 2 126 40027582
          BC106715.2 - 1, 2 126 40027584
          BC106716.1 - 1, 2 126 40027585
          DR001889.1 - 3, 4 253
          L38929.1 - 2, 3 228

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein dephosphorylation
          phosphate-containing compound metabolic process
          heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
          transmembrane receptor protein tyrosine phosphatase signaling pathway
          chemical synaptic transmission
          neuron differentiation
          peptidyl-tyrosine dephosphorylation
          positive regulation of dendrite morphogenesis
          presynaptic membrane assembly
          transmembrane receptor protein tyrosine phosphatase activity
          receptor binding
          protein binding
          cell adhesion molecule binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0