Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MMS19 SIRNA Assays ›
          Gene Symbol:
          MMS19
          Gene Name:
          MMS19 homolog, cytosolic iron-sulfur assembly component
          Gene Aliases:
          MET18, MMS19L, hMMS19
          Chromosome Location:
          Chr.10: 97458324 - 97498794 on Build GRCh38
          UniGene ID:
          Hs.500721
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 116327

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MMS19
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001289404.1 NP_001276333.1 5, 6 667
          NM_001289405.1 NP_001276334.1 7, 8 610
          NM_022362.4 NP_071757.4 6, 7 728
          XM_005270035.2 XP_005270092.1 5, 6 485
          XM_006717944.3 XP_006718007.2 6, 7 522
          XM_006717945.2 XP_006718008.1 6 949
          XM_011540063.2 XP_011538365.1 6 949
          XM_017016515.1 XP_016872004.1 7, 8 634
          XM_017016516.1 XP_016872005.1 7, 8 634
          XM_017016519.1 XP_016872008.1 7, 8 634
          XM_017016520.1 XP_016872009.1 6, 7 522
          XM_017016521.1 XP_016872010.1 6, 7 522
          XM_017016522.1 XP_016872011.1 6, 7 626
          XM_017016523.1 XP_016872012.1 5, 6 510
          XM_017016524.1 XP_016872013.1 5, 6 485
          XM_017016525.1 XP_016872014.1 5, 6 391
          XM_017016526.1 XP_016872015.1 5, 6 466
          XM_017016527.1 XP_016872016.1 5, 6 485
          XM_017016528.1 XP_016872017.1 6 949
          XM_017016529.1 XP_016872018.1 7 1017
          XM_017016530.1 XP_016872019.1 6, 7 626
          XM_017016531.1 XP_016872020.1 5, 6 485
          XM_017016533.1 XP_016872022.1 4, 5 342
          XM_017016534.1 XP_016872023.1 6 949
          XM_017016535.1 XP_016872024.1 6 949
          GenBank mRNA AF319947.1 - 7, 8 576
          AF357881.1 - 6, 7 527
          AJ306408.1 - 6, 7 510
          AK027710.1 - 5, 6 362
          AK056581.1 - 2, 3 125
          AK225312.1 - 6, 7 513
          AK298995.1 - 6, 7 566
          AK314374.1 - 6, 7 526
          BC080532.1 - 5, 6 446 4889876
          BC117129.1 - 6, 7 526 40125680

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorelay signal transduction system
          DNA metabolic process
          DNA repair
          nucleotide-excision repair
          transcription, DNA-templated
          cellular response to DNA damage stimulus
          chromosome segregation
          response to hormone
          iron-sulfur cluster assembly
          positive regulation of transcription, DNA-templated
          transcription coactivator activity
          protein binding
          enzyme binding
          receptor signaling complex scaffold activity
          estrogen receptor binding
          protein binding, bridging

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-8696d955b-mh7vc:80/100.66.135.11:80.
          git-commit: b6496d4449ebd8b46e03b29da6eff4841d0bfc13
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline