Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SLC5A11 SIRNA Assays ›
          Gene Symbol:
          SLC5A11
          Gene Name:
          solute carrier family 5 member 11
          Gene Aliases:
          KST1, RKST1, SGLT6, SMIT2
          Chromosome Location:
          Chr.16: 24832784 - 24911711 on Build GRCh38
          UniGene ID:
          Hs.164118
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 118939

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SLC5A11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001258411.2 NP_001245340.1 15 1943
          NM_001258412.2 NP_001245341.1 14 1838
          NM_001258413.2 NP_001245342.1 15 1889
          NM_001258414.1 NP_001245343.1 14 1613
          NM_052944.4 NP_443176.2 15 1899
          XM_005255080.2 XP_005255137.1 16 1927
          XM_005255082.4 XP_005255139.1 10 1333
          XM_006721015.2 XP_006721078.1 14 1907
          XM_011545725.2 XP_011544027.2 15 2070
          XM_011545728.1 XP_011544030.1 7 866
          XM_017022900.1 XP_016878389.1 16 2243
          XM_017022901.1 XP_016878390.1 16 2953
          XM_017022902.1 XP_016878391.1 16 2082
          XM_017022903.1 XP_016878392.1 17 2092
          XM_017022904.1 XP_016878393.1 17 2262
          XM_017022905.1 XP_016878394.1 15 2138
          XM_017022906.1 XP_016878395.1 16 2103
          XM_017022907.1 XP_016878396.1 15 1651
          XM_017022908.1 XP_016878397.1 15 1710
          XM_017022909.1 XP_016878398.1 14 1546
          GenBank mRNA AF292385.1 - 14 1667
          AJ305237.1 - 14 1685
          AK125267.1 - 17 2015
          AK295427.1 - 14 1707
          AK298373.1 - 14 1684
          AK303691.1 - 15 1811
          AY044906.1 - 15 1899
          BC049385.1 - 14 1483 5755223
          BC057780.1 - 15 1758 5287366

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          sodium ion transport
          apoptotic process
          carbohydrate transport
          polyol transport
          transmembrane transport
          protein binding
          polyol transmembrane transporter activity
          symporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0