Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143996.1 | NP_001137468.1 | 8 | 1025 | ||
NM_001143997.1 | NP_001137469.1 | 7 | 820 | |||
NM_018245.2 | NP_060715.2 | 9 | 1196 | |||
XM_011539946.1 | XP_011538248.1 | 9 | 1081 | |||
XM_011539947.1 | XP_011538249.1 | 9 | 1287 | |||
XM_017016401.1 | XP_016871890.1 | 9 | 1471 | |||
XM_017016402.1 | XP_016871891.1 | 9 | 1098 | |||
XM_017016403.1 | XP_016871892.1 | 7 | 844 | |||
GenBank mRNA | AB033116.3 | - | 8 | 1059 | ||
AK001713.1 | - | 9 | 1140 | |||
AK225558.1 | - | 9 | 1127 | |||
AK290226.1 | - | 8 | 1169 | |||
AK296551.1 | - | 8 | 934 | |||
AK303729.1 | - | 7 | 767 | |||
AK307759.1 | - | 8 | 955 | |||
BC026320.1 | - | 9 | 1115 | 4797814 |