Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GMEB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319674.1 | NP_001306603.1 | 7 | 758 | ||
NM_006582.3 | NP_006573.2 | 7 | 788 | |||
NM_024482.2 | NP_077808.1 | 7 | 755 | |||
XM_011540518.2 | XP_011538820.1 | 7 | 891 | |||
XM_011540519.2 | XP_011538821.1 | 7 | 731 | |||
XM_011540521.2 | XP_011538823.1 | 7 | 860 | |||
XM_011540522.2 | XP_011538824.1 | 7 | 707 | |||
XM_017000087.1 | XP_016855576.1 | 7 | 906 | |||
GenBank mRNA | AF099013.1 | - | 6 | 652 | ||
AF173868.1 | - | 7 | 683 | |||
AF203694.1 | - | 7 | 682 | |||
AK000892.1 | - | 7 | 754 | |||
AK023126.1 | - | 7 | 675 | |||
AK315195.1 | - | 7 | 734 | |||
AK315436.1 | - | 7 | 692 | |||
BC001473.2 | - | 7 | 705 | 3138481 |