Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020205.3 | NP_064590.2 | 7 | 1100 | ||
XM_011509782.1 | XP_011508084.1 | 7 | 1097 | |||
XM_011509783.2 | XP_011508085.1 | 7 | 1327 | |||
XM_011509784.2 | XP_011508086.1 | 7 | 891 | |||
XM_011509785.2 | XP_011508087.1 | 7 | 4524 | |||
XM_011509786.1 | XP_011508088.1 | 7 | 963 | |||
XM_011509787.1 | XP_011508089.1 | 7 | 706 | |||
XM_011509788.1 | XP_011508090.1 | 6 | 815 | |||
XM_017001850.1 | XP_016857339.1 | 7 | 963 | |||
XM_017001851.1 | XP_016857340.1 | 6 | 949 | |||
GenBank mRNA | AJ293573.1 | - | 7 | 943 | ||
AK299790.1 | - | 7 | 1086 | |||
BC020622.1 | - | 7 | 886 | |||
BC072681.1 | - | 7 | 1037 | 6170513 |