Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020205.3 | NP_064590.2 | 12 | 2923 | ||
XM_011509782.1 | XP_011508084.1 | 12 | 2947 | |||
XM_011509783.2 | XP_011508085.1 | 12 | 3177 | |||
XM_011509784.2 | XP_011508086.1 | 12 | 2741 | |||
XM_011509785.2 | XP_011508087.1 | 12 | 6374 | |||
XM_011509786.1 | XP_011508088.1 | 12 | 2813 | |||
XM_011509787.1 | XP_011508089.1 | 12 | 2556 | |||
XM_011509788.1 | XP_011508090.1 | 11 | 2665 | |||
XM_017001850.1 | XP_016857339.1 | 12 | 2786 | |||
XM_017001851.1 | XP_016857340.1 | 11 | 2772 | |||
GenBank mRNA | AJ293573.1 | - | 12 | 2766 | ||
AK299790.1 | - | 13 | 2822 | |||
AL122102.1 | - | 2 | 1260 | |||
BC072681.1 | - | 12 | 2860 | 6170513 |