Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTPN20 SIRNA Assays ›
          Gene Symbol:
          PTPN20
          Gene Name:
          protein tyrosine phosphatase, non-receptor type 20
          Gene Aliases:
          CT126, PTPN20A, PTPN20B, bA142I17.1, bA42B19.1
          Chromosome Location:
          Chr.10: 46911134 - 47003834 on Build GRCh38
          UniGene ID:
          Hs.440733
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 122821

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTPN20
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042357.4 NP_001035816.1 6 703
          NM_001042358.4 NP_001035817.1 5 561
          NM_001042359.3 NP_001035818.1 5 561
          NM_001042361.4 NP_001035820.1 5 561
          NM_001042363.4 NP_001035822.1 5 546
          NM_001042364.4 NP_001035823.1 4 451
          NM_001320681.1 NP_001307610.1 4 451
          NM_001320683.1 NP_001307612.1 6 718
          NM_001320684.1 NP_001307613.1 6 718
          NM_001320685.1 NP_001307614.1 5 623
          NM_001320686.1 NP_001307615.1 4 466
          NM_001320688.1 NP_001307617.1 4 466
          NM_001320691.1 NP_001307620.1 4 466
          NM_015605.8 NP_056420.3 6 703
          XM_011539605.2 XP_011537907.1 6 847
          XM_011539606.2 XP_011537908.1 6 1939
          XM_011539607.2 XP_011537909.1 6 838
          XM_011539610.2 XP_011537912.1 3 321
          XM_017016042.1 XP_016871531.1 6 682
          XM_017016043.1 XP_016871532.1 6 682
          XM_017016044.1 XP_016871533.1 5 604
          XM_017016046.1 XP_016871535.1 6 682
          XM_017016047.1 XP_016871536.1 6 682
          XM_017016048.1 XP_016871537.1 6 682
          XM_017016049.1 XP_016871538.1 4 413
          XM_017016050.1 XP_016871539.1 3 615
          XM_017016051.1 XP_016871540.1 6 682
          XM_017016052.1 XP_016871541.1 6 682
          XM_017016053.1 XP_016871542.1 6 682
          XM_017016054.1 XP_016871543.1 5 604
          XM_017016055.1 XP_016871544.1 6 682
          XM_017016056.1 XP_016871545.1 5 690
          XM_017016057.1 XP_016871546.1 4 449
          XM_017016058.1 XP_016871547.1 2 175
          XM_017016059.1 XP_016871548.1 4 593
          GenBank mRNA AK309922.1 - 5 765
          AL050040.1 - 4 409
          AY704141.1 - 5 502
          AY704142.1 - 5 502
          AY704144.1 - 5 502
          AY704146.1 - 5 551
          AY704147.1 - 6 659
          AY704148.1 - 6 659
          AY704150.1 - 5 564
          AY704151.1 - 4 456
          AY704152.1 - 4 407
          AY704153.1 - 4 407
          AY704154.1 - 4 407
          AY704158.1 - 4 407
          BC093750.1 - 5 557 7939595
          BC113473.1 - 5 557 8322525

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          peptidyl-tyrosine dephosphorylation
          protein tyrosine phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix