Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZCCHC17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282566.1 | NP_001269495.1 | 6 | 622 | ||
NM_001282567.1 | NP_001269496.1 | 6 | 562 | |||
NM_001282568.1 | NP_001269497.1 | 7 | 743 | |||
NM_001282569.1 | NP_001269498.1 | 8 | 675 | |||
NM_001282570.1 | NP_001269499.1 | 6 | 484 | |||
NM_001282571.1 | NP_001269500.1 | 6 | 504 | |||
NM_016505.3 | NP_057589.2 | 7 | 605 | |||
XM_006710681.3 | XP_006710744.1 | 8 | 749 | |||
XM_011541567.2 | XP_011539869.1 | 7 | 621 | |||
GenBank mRNA | AF151077.1 | - | 7 | 534 | ||
AF151085.1 | - | 7 | 561 | |||
AF247661.2 | - | 6 | 474 | |||
AK001929.1 | - | 7 | 555 | |||
AK022506.1 | - | 7 | 592 | |||
AK024049.1 | - | 7 | 581 | |||
AK302248.1 | - | 8 | 625 | |||
AK302343.1 | - | 6 | 508 | |||
AK307478.1 | - | 6 | 458 | |||
BC000685.2 | - | 7 | 643 | |||
BC007446.2 | - | 7 | 541 | 3009823 | ||
BC050609.1 | - | 7 | 545 | 6527025 | ||
BP309837.1 | - | 6 | 445 | |||
BP371153.1 | - | 6 | 440 |