Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHF21A SIRNA Assays ›
Gene Symbol:
PHF21A
Gene Name:
PHD finger protein 21A
Gene Aliases:
BHC80, BM-006
Chromosome Location:
Chr.11: 45929319 - 46121434 on Build GRCh38
UniGene ID:
Hs.502458
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
122941
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHF21A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001101802.1
NP_001095272.1
17, 18
2404
NM_016621.3
NP_057705.3
17, 18
2266
XM_005252965.4
XP_005253022.1
18, 19
2096
XM_011520157.2
XP_011518459.1
17, 18
1957
XM_011520158.2
XP_011518460.1
18, 19
2309
XM_011520160.2
XP_011518462.1
20, 21
2470
XM_011520161.2
XP_011518463.1
20, 21
2561
XM_011520162.2
XP_011518464.1
20, 21
2439
XM_011520164.2
XP_011518466.1
19, 20
2368
XM_011520165.2
XP_011518467.1
19, 20
2323
XM_011520166.2
XP_011518468.1
19, 20
2246
XM_011520167.2
XP_011518469.1
19, 20
2449
XM_011520168.2
XP_011518470.1
19, 20
2301
XM_011520169.2
XP_011518471.1
18, 19
2260
XM_011520173.2
XP_011518475.1
18, 19
2216
XM_011520174.2
XP_011518476.1
19, 20
2190
XM_011520175.2
XP_011518477.1
19, 20
2187
XM_011520179.2
XP_011518481.1
18, 19
2069
XM_017017885.1
XP_016873374.1
20, 21
2389
XM_017017886.1
XP_016873375.1
19, 20
2354
XM_017017887.1
XP_016873376.1
18, 19
2134
XM_017017888.1
XP_016873377.1
18, 19
2306
XM_017017889.1
XP_016873378.1
18, 19
2306
XM_017017890.1
XP_016873379.1
18, 19
2303
XM_017017891.1
XP_016873380.1
18, 19
2213
XM_017017892.1
XP_016873381.1
18, 19
2213
XM_017017894.1
XP_016873383.1
18, 19
2168
XM_017017895.1
XP_016873384.1
18, 19
2165
XM_017017896.1
XP_016873385.1
18, 19
2165
XM_017017897.1
XP_016873386.1
19, 20
2386
XM_017017898.1
XP_016873387.1
19, 20
2244
XM_017017899.1
XP_016873388.1
20, 21
2378
XM_017017900.1
XP_016873389.1
19, 20
2333
XM_017017901.1
XP_016873390.1
17, 18
2153
XM_017017902.1
XP_016873391.1
18, 19
2072
XM_017017903.1
XP_016873392.1
15, 16
1722
XM_017017905.1
XP_016873394.1
17, 18
2057
XM_017017907.1
XP_016873396.1
17, 18
1964
XM_017017908.1
XP_016873397.1
20, 21
2477
GenBank mRNA
AB051483.1
-
17, 18
2010
AF208848.1
-
9, 10
797
AK023258.1
-
19, 20
2027
AY090777.1
-
15, 16
1642
BC015714.2
-
18, 19
2068
3908900
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
blood coagulation
histone deacetylation
RNA polymerase II regulatory region sequence-specific DNA binding
chromatin binding
transcription cofactor activity
histone deacetylase activity
protein binding
zinc ion binding
histone binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0