Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017547.3 | NP_060017.1 | 2, 3 | 470 | ||
NR_037648.1 | 2, 3 | 489 | ||||
XM_011542895.2 | XP_011541197.1 | 2, 3 | 567 | |||
XM_011542896.2 | XP_011541198.1 | 2, 3 | 356 | |||
XM_017018000.1 | XP_016873489.1 | 2, 3 | 445 | |||
XM_017018001.1 | XP_016873490.1 | 2, 3 | 569 | |||
XM_017018003.1 | XP_016873492.1 | 2, 3 | 445 | |||
XM_017018004.1 | XP_016873493.1 | 2, 3 | 567 | |||
XM_017018006.1 | XP_016873495.1 | 2, 3 | 445 | |||
GenBank mRNA | AF103801.1 | - | 2, 3 | 368 | ||
AF447877.1 | - | 1, 2 | 717 | |||
AK023987.1 | - | 2, 3 | 339 | |||
AK295267.1 | - | 2, 3 | 425 | |||
AK295428.1 | - | 2, 3 | 339 | |||
AL136923.1 | - | 2, 3 | 350 | |||
BC013902.2 | - | 2, 3 | 326 | 3956972 | ||
DA056004.1 | - | 2, 3 | 470 |