Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MIER1 SIRNA Assays ›
          Gene Symbol:
          MIER1
          Gene Name:
          MIER1 transcriptional regulator
          Gene Aliases:
          ER1, MI-ER1
          Chromosome Location:
          Chr.1: 66924895 - 66988619 on Build GRCh38
          UniGene ID:
          Hs.605432
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 123256

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MIER1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077700.2 NP_001071168.2 6 752
          NM_001077701.2 NP_001071169.1 5 608
          NM_001077702.2 NP_001071170.2 7 826
          NM_001077703.2 NP_001071171.2 6 752
          NM_001077704.2 NP_001071172.1 5 608
          NM_001146110.1 NP_001139582.1 7 829
          NM_001146111.1 NP_001139583.1 7 826
          NM_001146112.1 NP_001139584.1 7 787
          NM_001146113.1 NP_001139585.1 5 606
          NM_020948.3 NP_065999.2 7 826
          XM_005271076.3 XP_005271133.1 7 864
          XM_011541864.2 XP_011540166.1 8 771
          XM_011541865.2 XP_011540167.1 8 732
          XM_017001919.1 XP_016857408.1 8 748
          XM_017001920.1 XP_016857409.1 8 765
          XM_017001921.1 XP_016857410.1 8 793
          XM_017001922.1 XP_016857411.1 8 768
          XM_017001923.1 XP_016857412.1 6 588
          XM_017001924.1 XP_016857413.1 7 720
          XM_017001925.1 XP_016857414.1 7 825
          XM_017001926.1 XP_016857415.1 8 764
          XM_017001927.1 XP_016857416.1 6 637
          XM_017001928.1 XP_016857417.1 6 713
          XM_017001929.1 XP_016857418.1 7 694
          XM_017001930.1 XP_016857419.1 6 638
          XM_017001931.1 XP_016857420.1 7 672
          XM_017001932.1 XP_016857421.1 5 601
          XM_017001933.1 XP_016857422.1 4 455
          XM_017001934.1 XP_016857423.1 3 351
          GenBank mRNA AB046830.1 - 3 377
          AF515446.1 - 5 608
          AF515447.1 - 6 689
          AF515448.2 - 7 763
          AK302061.1 - 6 752
          AK307717.1 - 7 723
          AL831987.2 - 7 768
          AY124186.2 - 7 763
          AY124187.1 - 6 689
          AY124188.1 - 5 608
          AY124189.2 - 7 763
          AY124190.1 - 6 689
          AY124191.1 - 5 608
          AY124192.2 - 7 763
          AY124193.1 - 6 689
          AY124194.1 - 5 608
          BC017423.1 - 5 585
          BC108726.1 - 7 707 5194296
          BC125217.1 - 4 404 40123797
          BC125218.1 - 4 404 40123803
          CR627441.1 - 7 663

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001131012, XM_008763881, XM_006238479, XM_008763879, XM_006238484, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001286222, NM_027696, NM_001286223, NM_001039081, NM_001286221, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          signal transduction
          positive regulation of chromatin silencing
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          DNA binding
          signal transducer activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-qp6lg:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0