Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM134 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078650.2 | NP_001072118.1 | 4 | 428 | ||
NM_001078651.2 | NP_001072119.1 | 4 | 401 | |||
NM_025124.3 | NP_079400.1 | 4 | 428 | |||
NR_073409.1 | 4 | 428 | ||||
NR_073410.1 | 4 | 412 | ||||
NR_073411.1 | 4 | 439 | ||||
NR_073412.1 | 4 | 428 | ||||
NR_073413.1 | 4 | 412 | ||||
XM_006718693.2 | XP_006718756.1 | 4 | 396 | |||
XM_011545268.2 | XP_011543570.1 | 4 | 423 | |||
XM_017018356.1 | XP_016873845.1 | 4 | 423 | |||
GenBank mRNA | AK025402.1 | - | 4 | 444 | ||
AK296988.1 | - | 4 | 357 | |||
AK303342.1 | - | 4 | 392 | |||
AK310743.1 | - | 4 | 316 | |||
AY007143.1 | - | 4 | 398 | |||
BC013883.1 | - | 4 | 397 | 3678954 | ||
BC125134.1 | - | 4 | 396 | 40119367 | ||
BG488634.1 | - | 4 | 350 | |||
BG719036.1 | - | 4 | 408 | |||
BM920758.1 | - | 396 | ||||
BX400047.2 | - | 4 | 384 | |||
CN336445.1 | - | 4 | 262 | |||
DN999439.1 | - | 4 | 417 | |||
HY053672.1 | - | 4 | 428 | |||
HY059123.1 | - | 4 | 355 |