Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145257.4 | NP_660300.3 | 3 | 549 | ||
GenBank mRNA | AK123465.1 | - | 2 | 338 | ||
AK291437.1 | - | 2 | 510 | |||
BC015419.1 | - | 2 | 523 | 4396825 | ||
BC060777.1 | - | 3 | 1454 | |||
BC071609.1 | - | 3 | 595 | 5264741 |