Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF101 SIRNA Assays ›
Gene Symbol:
C1orf101
Gene Name:
chromosome 1 open reading frame 101
Gene Aliases:
-
Chromosome Location:
Chr.1: 244452014 - 244640362 on Build GRCh38
UniGene ID:
Hs.459534
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
124241
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf101
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001130957.1
NP_001124429.1
4
308
NM_001242340.1
NP_001229269.1
3
259
NM_173807.4
NP_776168.1
4
308
XM_011544138.2
XP_011542440.1
4
359
XM_011544139.2
XP_011542441.1
3
254
XM_011544142.2
XP_011542444.1
5
582
XM_011544144.2
XP_011542446.1
3
259
XM_011544145.2
XP_011542447.1
3
231
XM_011544149.2
XP_011542451.1
4
244
XM_011544151.2
XP_011542453.1
4
275
XM_017000943.1
XP_016856432.1
4
360
XM_017000944.1
XP_016856433.1
3
738
XM_017000945.1
XP_016856434.1
4
361
XM_017000946.1
XP_016856435.1
3
459
XM_017000947.1
XP_016856436.1
4
360
XM_017000948.1
XP_016856437.1
4
253
XM_017000949.1
XP_016856438.1
4
534
XM_017000950.1
XP_016856439.1
4
258
XM_017000951.1
XP_016856440.1
4
360
XM_017000952.1
XP_016856441.1
4
360
XM_017000953.1
XP_016856442.1
4
361
GenBank mRNA
AK027205.1
-
5
498
AK302609.1
-
4
261
AK303059.1
-
3
219
BC032859.1
-
4
293
5269307
BX648685.1
-
3
219
DB462258.1
-
4
363
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.33.0-2025.07.30-1.0