Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB8OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308135.1 | NP_001295064.1 | 6 | 522 | ||
NM_001308136.1 | NP_001295065.1 | 6 | 490 | |||
NM_001308137.1 | NP_001295066.1 | 5 | 469 | |||
NM_001308138.1 | NP_001295067.1 | 6 | 587 | |||
NM_001308139.1 | NP_001295068.1 | 7 | 581 | |||
NM_001308140.1 | NP_001295069.1 | 7 | 574 | |||
NM_001308141.1 | NP_001295070.1 | 7 | 547 | |||
NM_178547.3 | NP_848642.1 | 7 | 543 | |||
XM_011541328.1 | XP_011539630.1 | 7 | 568 | |||
XM_011541329.2 | XP_011539631.1 | 7 | 468 | |||
XM_011541330.2 | XP_011539632.1 | 6 | 954 | |||
XM_011541334.2 | XP_011539636.1 | 6 | 913 | |||
XM_011541335.2 | XP_011539637.1 | 7 | 520 | |||
XM_011541337.2 | XP_011539639.1 | 7 | 555 | |||
XM_011541338.2 | XP_011539640.1 | 6 | 914 | |||
XM_017001134.1 | XP_016856623.1 | 7 | 554 | |||
XM_017001136.1 | XP_016856625.1 | 5 | 765 | |||
XM_017001137.1 | XP_016856626.1 | 7 | 402 | |||
XM_017001138.1 | XP_016856627.1 | 6 | 440 | |||
GenBank mRNA | AK289480.1 | - | 7 | 885 | ||
AY099494.1 | - | 7 | 443 | |||
AY151084.1 | - | 7 | 537 | |||
BC058843.1 | - | 6 | 460 | 6745279 | ||
BM465977.1 | - | 2 | 175 | |||
CN344731.1 | - | 6 | 430 | |||
DT220608.1 | - | 6 | 547 |