Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSTTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003082.1 | 1 | 174 | |||
GenBank mRNA | AK127991.1 | - | 1 | 174 |
Gene Symbol: GSTTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003081.1 | 1 | 82 | |||
GenBank mRNA | AL050257.1 | - | 1 | 82 | ||
BC096726.1 | - | 1 | 82 | |||
BC098108.1 | - | 1 | 82 | |||
BC098364.1 | - | 1 | 82 | |||
BC099710.1 | - | 1 | 82 |