Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YPEL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127399.1 | NP_001120871.1 | 4 | 712 | ||
NM_001127400.1 | NP_001120872.1 | 4 | 948 | |||
NM_001127401.1 | NP_001120873.1 | 5 | 1009 | |||
NM_016061.2 | NP_057145.1 | 3 | 651 | |||
XM_017004318.1 | XP_016859807.1 | 5 | 1247 | |||
XM_017004319.1 | XP_016859808.1 | 4 | 685 | |||
XM_017004320.1 | XP_016859809.1 | 4 | 1186 | |||
XM_017004321.1 | XP_016859810.1 | 4 | 878 | |||
XM_017004322.1 | XP_016859811.1 | 4 | 1109 | |||
GenBank mRNA | AF135161.1 | - | 3 | 555 | ||
AF151885.1 | - | 3 | 573 | |||
AK025730.1 | - | 4 | 589 | |||
AK307099.1 | - | 2 | 364 | |||
AK311404.1 | - | 3 | 362 | |||
BC000836.1 | - | 4 | 526 | 3456031 | ||
BC047237.1 | - | 4 | 1109 | 5269563 |