Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199803.2 | NP_001186732.1 | 2, 3 | 564 | ||
NM_001322101.1 | NP_001309030.1 | 3, 4 | 678 | |||
NM_024322.3 | NP_077298.1 | 3, 4 | 697 | |||
NR_136182.1 | 2, 3 | 583 | ||||
NR_136183.1 | 2, 3 | 583 | ||||
NR_136184.1 | 2, 3 | 564 | ||||
GenBank mRNA | AK027859.1 | - | 2, 3 | 298 | ||
AK315483.1 | - | 3, 4 | 323 | |||
AL514456.3 | - | 2, 3 | 257 | |||
BC002870.1 | - | 3, 4 | 354 | 3942844 | ||
BP366489.1 | - | 3, 4 | 350 | |||
BX406097.2 | - | 3, 4 | 353 | |||
CN263048.1 | - | 2, 3 | 220 | |||
CX872478.1 | - | 2, 3 | 201 |