Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM150A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031738.2 | NP_001026908.1 | 8 | 1511 | ||
NR_033179.1 | 7 | 1443 | ||||
XM_006711930.3 | XP_006711993.1 | 8 | 1711 | |||
XM_006711931.3 | XP_006711994.1 | 8 | 1550 | |||
XM_006711932.2 | XP_006711995.1 | 6 | 1680 | |||
XM_011532505.2 | XP_011530807.1 | 7 | 1546 | |||
XM_011532507.2 | XP_011530809.1 | 7 | 2335 | |||
XM_011532508.2 | XP_011530810.1 | 6 | 1478 | |||
XM_017003305.1 | XP_016858794.1 | 7 | 2367 | |||
XM_017003306.1 | XP_016858795.1 | 8 | 2100 | |||
XM_017003307.1 | XP_016858796.1 | 7 | 2166 | |||
GenBank mRNA | AK074505.1 | - | 7 | 1444 | ||
AK098152.1 | - | 5 | 2282 | |||
BC050466.1 | - | 8 | 1471 | 6197700 | ||
BX647409.1 | - | 7 | 1379 |