Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 3 | 1522 | ||
NM_001321178.1 | NP_001308107.1 | 3 | 3288 | |||
NM_001321179.1 | NP_001308108.1 | 4 | 2547 | |||
NM_001321180.1 | NP_001308109.1 | 4 | 1685 | |||
NM_001321181.1 | NP_001308110.1 | 4 | 1657 | |||
NM_001321182.1 | NP_001308111.1 | 3 | 1546 | |||
NM_001321183.1 | NP_001308112.1 | 4 | 1366 | |||
NM_001321186.1 | NP_001308115.1 | 4 | 1248 | |||
NM_001321187.1 | NP_001308116.1 | 4 | 1230 | |||
NM_001321188.1 | NP_001308117.1 | 4 | 1174 | |||
XM_011530817.1 | XP_011529119.1 | 4 | 1639 | |||
XM_011530821.2 | XP_011529123.1 | 3 | 1147 | |||
GenBank mRNA | AK091598.1 | - | 4 | 2443 | ||
BX649013.1 | - | 3 | 3251 |