Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM156A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242489.1 | NP_001229418.1 | 3 | 1847 | ||
NM_001242490.1 | NP_001229419.1 | 4 | 1831 | |||
NM_001242491.1 | NP_001229420.1 | 3 | 1794 | |||
NM_001242492.1 | NP_001229421.1 | 5 | 1680 | |||
NM_001242493.1 | NP_001229422.1 | 5 | 1573 | |||
NM_001242494.1 | NP_001229423.1 | 4 | 1531 | |||
NM_001242495.1 | NP_001229424.1 | 3 | 1438 | |||
NM_001242496.1 | NP_001229425.1 | 4 | 1422 | |||
NM_001242497.1 | NP_001229426.1 | 3 | 1329 | |||
NM_014138.4 | NP_054857.2 | 5 | 1982 | |||
XM_006724586.3 | XP_006724649.1 | 4 | 3079 | |||
XM_006724587.3 | XP_006724650.1 | 4 | 2670 | |||
XM_011530781.1 | XP_011529083.1 | 4 | 1887 | |||
XM_011530782.1 | XP_011529084.1 | 4 | 2156 | |||
XM_011530783.1 | XP_011529085.1 | 4 | 1614 | |||
XM_011530784.2 | XP_011529086.1 | 3 | 1383 | |||
XM_017029457.1 | XP_016884946.1 | 5 | 3838 | |||
XM_017029458.1 | XP_016884947.1 | 5 | 3429 | |||
GenBank mRNA | AF090943.1 | - | 1 | 1022 | ||
AF370413.1 | - | 3 | 1998 | |||
AK054721.1 | - | 3 | 2408 | |||
AK057692.1 | - | 3 | 1777 | |||
AK092365.1 | - | 4 | 2126 | |||
AK096162.1 | - | 3 | 2282 | |||
AL834135.1 | - | 3 | 1446 | |||
BC000867.1 | - | 1 | 997 | 3460178 | ||
BC067819.1 | - | 3 | 1430 | 5313567 |
Gene Symbol: FAM156B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099684.2 | NP_001093154.1 | 3 | 1770 | ||
NM_001321178.1 | NP_001308107.1 | 3 | 3536 | |||
NM_001321179.1 | NP_001308108.1 | 4 | 2795 | |||
NM_001321180.1 | NP_001308109.1 | 4 | 1933 | |||
NM_001321181.1 | NP_001308110.1 | 4 | 1905 | |||
NM_001321182.1 | NP_001308111.1 | 3 | 1794 | |||
NM_001321183.1 | NP_001308112.1 | 4 | 1614 | |||
NM_001321186.1 | NP_001308115.1 | 4 | 1496 | |||
NM_001321187.1 | NP_001308116.1 | 4 | 1478 | |||
NM_001321188.1 | NP_001308117.1 | 4 | 1422 | |||
XM_011530817.1 | XP_011529119.1 | 4 | 1887 | |||
XM_011530821.2 | XP_011529123.1 | 3 | 1395 | |||
GenBank mRNA | AK091598.1 | - | 4 | 2691 | ||
BX649013.1 | - | 3 | 3499 |