Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018268.3 | NP_060738.2 | 10, 11 | 1332 | ||
XM_005248551.4 | XP_005248608.1 | 8, 9 | 1125 | |||
XM_005248552.3 | XP_005248609.1 | 10, 11 | 1299 | |||
XM_011543505.2 | XP_011541807.1 | 10, 11 | 1094 | |||
XM_017009606.1 | XP_016865095.1 | 10, 11 | 1742 | |||
GenBank mRNA | AF115511.1 | - | 10, 11 | 1080 | ||
AK001766.1 | - | 10, 11 | 1278 | |||
AK021855.1 | - | 10, 11 | 1118 | |||
AK023977.1 | - | 10, 11 | 1034 | |||
AK293316.1 | - | 8, 9 | 817 | |||
AK295249.1 | - | 9, 10 | 939 | |||
AK300058.1 | - | 8, 9 | 943 | |||
AK304329.1 | - | 10, 11 | 1024 | |||
AL834138.1 | - | 4, 5 | 2013 | |||
BC040241.1 | - | 10, 11 | 1084 | 3447269 | ||
KU178690.1 | - | 10, 11 | 1012 | |||
KU178691.1 | - | 10, 11 | 1027 |