Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC136 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022742.4 | NP_073579.4 | 4 | 1052 | ||
XM_005250534.3 | XP_005250591.1 | 5 | 836 | |||
XM_005250538.1 | XP_005250595.1 | 5 | 836 | |||
XM_011516483.1 | XP_011514785.1 | 5 | 836 | |||
XM_011516484.1 | XP_011514786.1 | 5 | 836 | |||
XM_011516485.1 | XP_011514787.1 | 5 | 836 | |||
XM_011516487.2 | XP_011514789.1 | 5 | 836 | |||
XM_011516488.2 | XP_011514790.1 | 5 | 836 | |||
XM_011516489.1 | XP_011514791.1 | 5 | 836 | |||
XM_011516490.2 | XP_011514792.1 | 5 | 836 | |||
XM_017012533.1 | XP_016868022.1 | 5 | 1302 | |||
XM_017012535.1 | XP_016868024.1 | 5 | 836 | |||
GenBank mRNA | AB058696.1 | - | 4 | 1003 | ||
AL133027.1 | - | 1 | 266 | |||
BC070114.1 | - | 1 | 406 | |||
BC150331.1 | - | 4 | 1014 | 8860498 | ||
BC152408.1 | - | 4 | 1003 | 8862511 |