Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C5ORF42 SIRNA Assays ›
Gene Symbol:
C5orf42
Gene Name:
chromosome 5 open reading frame 42
Gene Aliases:
Hug, JBTS17, OFD6
Chromosome Location:
Chr.5: 37063928 - 37249428 on Build GRCh38
UniGene ID:
Hs.586199
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
127348
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C5orf42
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_023073.3
NP_075561.3
26
5171
XM_005248345.3
XP_005248402.1
26
5159
XM_005248346.3
XP_005248403.1
26
5079
XM_005248347.3
XP_005248404.1
26
5079
XM_005248349.3
XP_005248406.1
26
5079
XM_005248350.3
XP_005248407.1
26
4948
XM_011514085.2
XP_011512387.1
26
5078
XM_011514086.2
XP_011512388.1
26
5075
XM_011514087.2
XP_011512389.1
26
5164
XM_011514088.2
XP_011512390.1
26
5164
XM_011514089.2
XP_011512391.1
26
5164
XM_011514090.2
XP_011512392.1
26
4942
XM_011514092.2
XP_011512394.1
26
5164
XM_011514094.2
XP_011512396.1
11
2213
XM_017009760.1
XP_016865249.1
25
4890
XM_017009761.1
XP_016865250.1
25
4858
XM_017009762.1
XP_016865251.1
22
4617
XM_017009763.1
XP_016865252.1
22
4490
XM_017009764.1
XP_016865253.1
22
4451
XM_017009765.1
XP_016865254.1
26
4859
XM_017009766.1
XP_016865255.1
12
2341
GenBank mRNA
AK055192.1
-
2
516
AL833065.2
-
1
406
BC144069.1
-
11
2084
9052589
BC144070.1
-
11
1955
BC150594.1
-
11
2084
9056964
Back To Top
More Information
Gene Ontology Categories:
Function(s)
establishment of planar polarity
kidney development
ventricular septum development
cerebellum development
cilium assembly
embryonic digit morphogenesis
palate development
coronary vasculature development
protein localization to ciliary transition zone
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix