Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM185A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145268.1 | NP_001138740.1 | 6 | 1123 | ||
NM_001145269.1 | NP_001138741.1 | 5 | 772 | |||
NR_026879.1 | 6 | 882 | ||||
XM_006715896.3 | XP_006715959.1 | 6 | 1129 | |||
XM_011515922.2 | XP_011514224.1 | 6 | 1130 | |||
XM_011515923.2 | XP_011514225.1 | 6 | 1112 | |||
XM_011515924.2 | XP_011514226.1 | 6 | 1130 | |||
XM_011515925.2 | XP_011514227.1 | 5 | 1088 | |||
XM_011515926.2 | XP_011514228.1 | 6 | 1130 | |||
XM_017011846.1 | XP_016867335.1 | 6 | 1130 | |||
XM_017011847.1 | XP_016867336.1 | 5 | 1088 | |||
XM_017011848.1 | XP_016867337.1 | 6 | 1130 | |||
XM_017011849.1 | XP_016867338.1 | 5 | 1070 | |||
GenBank mRNA | AK054745.1 | - | 6 | 792 | ||
AK297455.1 | - | 7 | 603 | |||
AK298753.1 | - | 5 | 696 | |||
BC029175.1 | - | 6 | 1082 |