Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4orf33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099783.1 | NP_001093253.1 | 6 | 936 | ||
NM_173487.2 | NP_775758.2 | 6 | 1208 | |||
XM_005262734.4 | XP_005262791.1 | 6 | 1203 | |||
GenBank mRNA | AK091022.1 | - | 6 | 1197 | ||
BC016358.2 | - | 2 | 549 | |||
BC032582.1 | - | 6 | 1139 | 5494700 | ||
BX538164.1 | - | 6 | 1149 |