Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RALYL SIRNA Assays ›
          Gene Symbol:
          RALYL
          Gene Name:
          RALY RNA binding protein-like
          Gene Aliases:
          HNRPCL3
          Chromosome Location:
          Chr.8: 84182629 - 84921844 on Build GRCh38
          UniGene ID:
          Hs.121663
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 128319

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RALYL
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100391.1 NP_001093861.1 6 710
          NM_001100392.1 NP_001093862.1 7 953
          NM_001100393.1 NP_001093863.1 6 921
          NM_001287243.1 NP_001274172.1 5 1601
          NM_001287244.1 NP_001274173.1 5 415
          NM_173848.5 NP_776247.3 6 1634
          XM_011517462.2 XP_011515764.1 6 1705
          XM_017013058.1 XP_016868547.1 5 876
          XM_017013059.1 XP_016868548.1 8 1004
          XM_017013060.1 XP_016868549.1 6 3154
          XM_017013061.1 XP_016868550.1 6 2306
          XM_017013062.1 XP_016868551.1 8 1113
          XM_017013063.1 XP_016868552.1 6 876
          XM_017013064.1 XP_016868553.1 7 1227
          XM_017013065.1 XP_016868554.1 6 775
          XM_017013066.1 XP_016868555.1 5 888
          XM_017013067.1 XP_016868556.1 6 1779
          XM_017013068.1 XP_016868557.1 7 971
          XM_017013069.1 XP_016868558.1 7 1027
          XM_017013070.1 XP_016868559.1 6 871
          XM_017013071.1 XP_016868560.1 5 742
          XM_017013072.1 XP_016868561.1 6 862
          XM_017013073.1 XP_016868562.1 6 862
          XM_017013074.1 XP_016868563.1 6 775
          XM_017013075.1 XP_016868564.1 5 827
          XM_017013076.1 XP_016868565.1 6 655
          XM_017013077.1 XP_016868566.1 7 1905
          XM_017013078.1 XP_016868567.1 6 921
          XM_017013079.1 XP_016868568.1 7 904
          XM_017013080.1 XP_016868569.1 5 622
          XM_017013081.1 XP_016868570.1 5 622
          XM_017013082.1 XP_016868571.1 5 888
          XM_017013083.1 XP_016868572.1 5 576
          XM_017013084.1 XP_016868573.1 4 1411
          XM_017013085.1 XP_016868574.1 6 458
          XM_017013086.1 XP_016868575.1 5 415
          XM_017013087.1 XP_016868576.1 5 415
          GenBank mRNA AF495529.1 - 6 1634
          AF495530.1 - 6 851
          AK094543.1 - 6 865
          AK095026.1 - 5 1417
          AK095560.1 - 7 904
          AK123437.1 - 5 415
          BC031090.1 - 6 699 5311935

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          nucleotide binding
          protein binding
          identical protein binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline