Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMSAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015447.3 | NP_056262.3 | 17 | 6072 | ||
XM_005263396.3 | XP_005263453.1 | 18 | 6497 | |||
XM_005263397.1 | XP_005263454.1 | 15 | 5598 | |||
XM_011518272.1 | XP_011516574.1 | 17 | 5766 | |||
XM_011518273.2 | XP_011516575.1 | 16 | 5597 | |||
XM_011518274.1 | XP_011516576.1 | 15 | 5627 | |||
XM_017014301.1 | XP_016869790.1 | 17 | 6353 | |||
XM_017014302.1 | XP_016869791.1 | 17 | 6277 | |||
GenBank mRNA | AJ420438.1 | - | 1 | 552 | ||
AJ519841.1 | - | 15 | 5600 | |||
AK025533.1 | - | 1 | 508 | |||
AK055790.1 | - | 1 | 1034 | |||
AL834528.1 | - | 7 | 2424 | |||
BC010646.1 | - | 7 | 2798 | |||
BC012778.2 | - | 7 | 4072 |