Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KLHL32 SIRNA Assays ›
Gene Symbol:
KLHL32
Gene Name:
kelch like family member 32
Gene Aliases:
BKLHD5, KIAA1900, UG0030H05, dJ21F7.1
Chromosome Location:
Chr.6: 96898083 - 97142949 on Build GRCh38
UniGene ID:
Hs.45056
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
128869
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KLHL32
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286252.1
NP_001273181.1
4
718
NM_001286254.2
NP_001273183.1
2
232
NM_001323252.1
NP_001310181.1
5
765
NM_001323253.1
NP_001310182.1
4
718
NM_001323254.1
NP_001310183.1
4
718
NM_001323255.1
NP_001310184.1
4
718
NM_001323256.1
NP_001310185.1
4
718
NM_001323257.1
NP_001310186.1
4
718
NM_001323258.1
NP_001310187.1
5
2462
NM_001323259.1
NP_001310188.1
4
718
NM_001323260.1
NP_001310189.1
4
718
NM_001323261.1
NP_001310190.1
2
232
NM_001323262.1
NP_001310191.1
2
232
NM_001323263.1
NP_001310192.1
2
232
NM_001323264.1
NP_001310193.1
2
232
NM_052904.3
NP_443136.2
4
718
NR_104421.1
4
718
NR_136579.1
2
232
NR_136580.1
4
718
XM_005266813.4
XP_005266870.1
2
123
XM_017010228.1
XP_016865717.1
2
123
GenBank mRNA
AB067487.1
-
2
132
AK055292.1
-
4
604
BC031830.1
-
3
299
BC047491.1
-
2
112
BC071627.1
-
4
593
5245540
BI601157.1
-
2
118
BX645412.1
-
2
239
DA332224.1
-
5
490
HY131926.1
-
2
114
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
ubiquitin-protein transferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix