Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BEND7 SIRNA Assays ›
          Gene Symbol:
          BEND7
          Gene Name:
          BEN domain containing 7
          Gene Aliases:
          C10orf30
          Chromosome Location:
          Chr.10: 13438484 - 13528772 on Build GRCh38
          UniGene ID:
          Hs.498740
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 129566

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BEND7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001100912.1 NP_001094382.1 6 1158
          NM_152751.2 NP_689964.2 7 1289
          XM_005252402.4 XP_005252459.1 7 1387
          XM_011519390.2 XP_011517692.1 6 1080
          XM_011519391.2 XP_011517693.1 6 1080
          XM_011519392.2 XP_011517694.1 7 1386
          XM_011519393.2 XP_011517695.1 6 1175
          XM_011519394.2 XP_011517696.1 8 1322
          XM_011519395.2 XP_011517697.1 6 1682
          XM_011519396.2 XP_011517698.1 7 1773
          XM_011519397.2 XP_011517699.1 7 1328
          XM_011519398.2 XP_011517700.1 6 1080
          XM_011519399.2 XP_011517701.1 6 1080
          XM_011519400.2 XP_011517702.1 6 1080
          XM_011519401.2 XP_011517703.1 6 1080
          XM_011519402.2 XP_011517704.1 6 1080
          XM_011519403.2 XP_011517705.1 6 1080
          XM_011519404.2 XP_011517706.1 6 1080
          XM_011519405.2 XP_011517707.1 6 1080
          XM_011519406.2 XP_011517708.1 6 1080
          XM_011519407.1 XP_011517709.1 6 1080
          XM_011519408.2 XP_011517710.1 6 1080
          XM_011519410.2 XP_011517712.1 6 1080
          XM_011519411.2 XP_011517713.1 6 1080
          XM_011519413.1 XP_011517715.1 5 854
          XM_011519414.1 XP_011517716.1 5 854
          XM_017015915.1 XP_016871404.1 8 1307
          XM_017015916.1 XP_016871405.1 6 1080
          XM_017015917.1 XP_016871406.1 6 1080
          XM_017015918.1 XP_016871407.1 7 1321
          XM_017015919.1 XP_016871408.1 7 1343
          GenBank mRNA AK097602.1 - 7 1289
          AL832195.1 - 2 260
          BC031618.1 - 6 1127 5172600

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-z8v7b:80/100.66.130.62:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0