Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164674.1 | NP_001158146.1 | 7 | 961 | ||
NM_182760.3 | NP_877437.2 | 8 | 1036 | |||
XM_011533624.2 | XP_011531926.1 | 8 | 1027 | |||
XM_011533625.2 | XP_011531927.1 | 8 | 1027 | |||
XM_011533626.2 | XP_011531928.1 | 8 | 1027 | |||
XM_017006253.1 | XP_016861742.1 | 7 | 949 | |||
XM_017006254.1 | XP_016861743.1 | 8 | 1027 | |||
XM_017006255.1 | XP_016861744.1 | 8 | 1027 | |||
GenBank mRNA | AB448737.1 | - | 8 | 1007 | ||
AK057983.1 | - | 9 | 935 | |||
AK075459.1 | - | 8 | 1018 | |||
AK295240.1 | - | 4 | 627 | |||
AK302018.1 | - | 7 | 929 | |||
AY208752.1 | - | 8 | 999 | |||
AY323910.1 | - | 8 | 999 | |||
AY358092.1 | - | 8 | 1016 | |||
BC017005.2 | - | 3 | 275 | |||
BC110862.1 | - | 8 | 1007 | 5760490 | ||
BC121122.2 | - | 8 | 1028 | 40120670 |