Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WNK2 SIRNA Assays ›
          Gene Symbol:
          WNK2
          Gene Name:
          WNK lysine deficient protein kinase 2
          Gene Aliases:
          NY-CO-43, P/OKcl.13, PRKWNK2, SDCCAG43
          Chromosome Location:
          Chr.9: 93184156 - 93327581 on Build GRCh38
          UniGene ID:
          Hs.654856
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 130209

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WNK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001282394.1 NP_001269323.1 5, 6 1325
          NM_006648.3 NP_006639.3 5, 6 1325
          XM_005252137.3 XP_005252194.1 6, 7 1557
          XM_005252140.2 XP_005252197.1 6, 7 1557
          XM_005252141.2 XP_005252198.1 6, 7 1557
          XM_005252144.2 XP_005252201.1 6, 7 1557
          XM_011518926.2 XP_011517228.1 6, 7 1557
          XM_011518927.2 XP_011517229.1 6, 7 1557
          XM_011518928.2 XP_011517230.1 6, 7 1557
          XM_011518929.2 XP_011517231.1 6, 7 1557
          XM_011518930.2 XP_011517232.1 6, 7 1557
          XM_011518931.2 XP_011517233.1 6, 7 1557
          XM_011518932.2 XP_011517234.1 6, 7 1557
          XM_011518935.2 XP_011517237.1 6, 7 1557
          XM_011518936.2 XP_011517238.1 6, 7 1557
          XM_011518937.2 XP_011517239.1 5, 6 865
          XM_011518939.2 XP_011517241.1 5, 6 910
          XM_011518940.2 XP_011517242.1 5, 6 912
          XM_017015045.1 XP_016870534.1 6, 7 1557
          XM_017015046.1 XP_016870535.1 6, 7 1557
          XM_017015047.1 XP_016870536.1 6, 7 1557
          XM_017015048.1 XP_016870537.1 6, 7 1557
          XM_017015049.1 XP_016870538.1 6, 7 1557
          XM_017015050.1 XP_016870539.1 6, 7 1557
          XM_017015051.1 XP_016870540.1 6, 7 1557
          XM_017015052.1 XP_016870541.1 6, 7 1557
          XM_017015053.1 XP_016870542.1 6, 7 1557
          XM_017015054.1 XP_016870543.1 6, 7 1557
          XM_017015055.1 XP_016870544.1 6, 7 1557
          GenBank mRNA AB044546.1 - 5, 6 1349
          AB051547.2 - 5, 6 1312
          AJ242724.4 - 5, 6 1325
          AK310390.1 - 6, 7 750

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          negative regulation of cell proliferation
          intracellular signal transduction
          protein autophosphorylation
          negative regulation of ERK1 and ERK2 cascade
          regulation of ion homeostasis
          positive regulation of sodium ion transmembrane transporter activity
          protein serine/threonine kinase activity
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0