Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ZDHHC11 SIRNA Assays ›
          Gene Symbol:
          ZDHHC11 ZDHHC11B
          Gene Name:
          zinc finger DHHC-type containing 11
          zinc finger DHHC-type containing 11B
          Gene Aliases:
          ZNF399
          DHHC-11B
          Chromosome Location:
          Chr.5: 795605 - 863634 on Build GRCh38
          Chromosome Location:
          Chr.5: 710355 - 784749 on Build GRCh38
          UniGene ID:
          Hs.654874
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 130726

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ZDHHC11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_024786.2 NP_079062.1 3, 4 883
          XM_017009869.1 XP_016865358.1 4, 5 764
          XM_017009870.1 XP_016865359.1 6, 7 2575
          XM_017009871.1 XP_016865360.1 6, 7 2524
          XM_017009872.1 XP_016865361.1 4, 5 599
          GenBank mRNA AK023215.1 - 3, 4 883
          BC032000.1 - 3, 4 502 4750082
          Gene Symbol: ZDHHC11B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017010110.1 XP_016865599.1 7, 8 1148
          XM_017010111.1 XP_016865600.1 6, 7 975
          XM_017010112.1 XP_016865601.1 6, 7 1552
          XM_017010113.1 XP_016865602.1 7, 8 1674
          XM_017010114.1 XP_016865603.1 7, 8 4435
          XM_017010115.1 XP_016865604.1 6, 7 1500
          XM_017010116.1 XP_016865605.1 5, 6 1900
          XM_017010117.1 XP_016865606.1 5, 6 1735
          XM_017010118.1 XP_016865607.1 5, 6 1900
          XM_017010119.1 XP_016865608.1 5, 6 1900
          XM_017010120.1 XP_016865609.1 5, 6 1900
          XM_017010121.1 XP_016865610.1 5, 6 1900
          XM_017010122.1 XP_016865611.1 5, 6 1900
          XM_017010123.1 XP_016865612.1 5, 6 1649
          XM_017010124.1 XP_016865613.1 6, 7 1659
          XM_017010125.1 XP_016865614.1 2, 3 1224

          Back To Top

          More Information


          Gene Ontology Categories:

          Process(es)

          zinc ion binding
          protein-cysteine S-palmitoyltransferase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0