Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK11 SIRNA Assays ›
Gene Symbol:
NEK11
Gene Name:
NIMA related kinase 11
Gene Aliases:
-
Chromosome Location:
Chr.3: 131026850 - 131350465 on Build GRCh38
UniGene ID:
Hs.657336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130736
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146003.1
NP_001139475.1
2, 3
418
NM_001321220.1
NP_001308149.1
2, 3
418
NM_001321221.1
NP_001308150.1
3, 4
491
NM_001321222.1
NP_001308151.1
2, 3
418
NM_001321223.1
NP_001308152.1
2, 3
536
NM_024800.4
NP_079076.3
3, 4
491
NM_145910.3
NP_665917.1
3, 4
491
XM_011513167.2
XP_011511469.1
3, 4
691
XM_017007205.1
XP_016862694.1
3, 4
668
XM_017007206.1
XP_016862695.1
3, 4
586
XM_017007207.1
XP_016862696.1
3, 4
802
XM_017007208.1
XP_016862697.1
2, 3
386
XM_017007209.1
XP_016862698.1
2, 3
595
XM_017007210.1
XP_016862699.1
2, 3
451
XM_017007211.1
XP_016862700.1
2, 3
386
XM_017007212.1
XP_016862701.1
3, 4
490
XM_017007213.1
XP_016862702.1
3, 4
490
XM_017007214.1
XP_016862703.1
2, 3
593
XM_017007215.1
XP_016862704.1
3, 4
490
XM_017007216.1
XP_016862705.1
2, 3
386
XM_017007217.1
XP_016862706.1
3, 4
490
XM_017007220.1
XP_016862709.1
2, 3
386
XM_017007221.1
XP_016862710.1
2, 3
386
XM_017007222.1
XP_016862711.1
2, 3
386
XM_017007223.1
XP_016862712.1
2, 3
386
XM_017007224.1
XP_016862713.1
3, 4
408
XM_017007229.1
XP_016862718.1
3, 4
490
XM_017007230.1
XP_016862719.1
3, 4
668
XM_017007233.1
XP_016862722.1
1, 2
98
GenBank mRNA
AB071996.1
-
3, 4
458
AB071997.1
-
3, 4
458
AK123021.1
-
2, 3
1385
AK297300.1
-
2, 3
418
AK316352.1
-
2, 3
333
AL833472.1
-
2, 3
389
BC028587.2
-
2, 3
535
4829802
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
histone phosphorylation
G2 DNA damage checkpoint
intra-S DNA damage checkpoint
intracellular signal transduction
mitotic cell cycle phase transition
regulation of mitotic cell cycle phase transition
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0