Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NEK11 SIRNA Assays ›
Gene Symbol:
NEK11
Gene Name:
NIMA related kinase 11
Gene Aliases:
-
Chromosome Location:
Chr.3: 131026850 - 131350465 on Build GRCh38
UniGene ID:
Hs.657336
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
130738
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NEK11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146003.1
NP_001139475.1
9
1200
NM_001321220.1
NP_001308149.1
9
1200
NM_001321221.1
NP_001308150.1
11
1399
NM_001321223.1
NP_001308152.1
9
1318
NM_001321224.1
NP_001308153.1
8
1034
NM_024800.4
NP_079076.3
10
1273
NM_145910.3
NP_665917.1
10
1273
XM_011513167.2
XP_011511469.1
10
1473
XM_011513173.1
XP_011511475.1
8
949
XM_011513174.1
XP_011511476.1
10
1219
XM_011513175.1
XP_011511477.1
9
1061
XM_011513181.1
XP_011511483.1
6
602
XM_017007205.1
XP_016862694.1
11
1576
XM_017007206.1
XP_016862695.1
11
1494
XM_017007207.1
XP_016862696.1
11
1710
XM_017007208.1
XP_016862697.1
10
1294
XM_017007209.1
XP_016862698.1
10
1503
XM_017007210.1
XP_016862699.1
10
1359
XM_017007211.1
XP_016862700.1
10
1294
XM_017007212.1
XP_016862701.1
10
1272
XM_017007213.1
XP_016862702.1
10
1272
XM_017007214.1
XP_016862703.1
9
1375
XM_017007215.1
XP_016862704.1
10
1272
XM_017007216.1
XP_016862705.1
9
1168
XM_017007218.1
XP_016862707.1
12
1477
XM_017007219.1
XP_016862708.1
9
971
XM_017007220.1
XP_016862709.1
10
1294
XM_017007221.1
XP_016862710.1
10
1294
XM_017007222.1
XP_016862711.1
10
1294
XM_017007224.1
XP_016862713.1
11
1338
XM_017007225.1
XP_016862714.1
11
1351
XM_017007226.1
XP_016862715.1
11
1615
XM_017007227.1
XP_016862716.1
9
1095
XM_017007228.1
XP_016862717.1
7
710
XM_017007231.1
XP_016862720.1
9
1073
XM_017007232.1
XP_016862721.1
10
1278
XM_017007233.1
XP_016862722.1
9
1028
GenBank mRNA
AB071996.1
-
10
1240
AB071997.1
-
10
1240
AK027148.1
-
5
466
AK293261.1
-
8
949
AL833472.1
-
9
1171
BC028587.2
-
9
1317
4829802
BX328072.2
-
3
268
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
histone phosphorylation
G2 DNA damage checkpoint
intra-S DNA damage checkpoint
intracellular signal transduction
mitotic cell cycle phase transition
regulation of mitotic cell cycle phase transition
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.31.0-2025.06.23-1.0