Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FANK1 SIRNA Assays ›
          Gene Symbol:
          FANK1
          Gene Name:
          fibronectin type III and ankyrin repeat domains 1
          Gene Aliases:
          HSD13
          Chromosome Location:
          Chr.10: 125896539 - 126009599 on Build GRCh38
          UniGene ID:
          Hs.352591
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 131799

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FANK1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_145235.3 NP_660278.3 5, 6 585
          XM_005270280.3 XP_005270337.1 4, 5 401
          XM_006718058.3 XP_006718121.1 7, 8 584
          XM_006718064.3 XP_006718127.1 6, 7 578
          XM_011540348.2 XP_011538650.1 7, 8 699
          XM_011540349.2 XP_011538651.1 7, 8 703
          XM_011540350.2 XP_011538652.1 6, 7 662
          XM_011540351.2 XP_011538653.1 6, 7 638
          XM_011540356.2 XP_011538658.1 5, 6 550
          XM_011540357.2 XP_011538659.1 7, 8 646
          XM_011540358.2 XP_011538660.1 5, 6 565
          XM_011540360.2 XP_011538662.1 5, 6 479
          XM_011540361.2 XP_011538663.1 5, 6 414
          XM_011540362.2 XP_011538664.1 3, 4 300
          XM_017016916.1 XP_016872405.1 7, 8 645
          XM_017016917.1 XP_016872406.1 7, 8 780
          XM_017016918.1 XP_016872407.1 6, 7 587
          XM_017016919.1 XP_016872408.1 6, 7 739
          XM_017016921.1 XP_016872410.1 6, 7 702
          XM_017016922.1 XP_016872411.1 6, 7 602
          XM_017016923.1 XP_016872412.1 5, 6 584
          XM_017016924.1 XP_016872413.1 8, 9 757
          XM_017016925.1 XP_016872414.1 4, 5 7307
          XM_017016926.1 XP_016872415.1 2, 3 714
          GenBank mRNA AY251163.1 - 4, 5 485
          AY358154.1 - 6, 7 641
          BC024189.2 - 5, 6 585 4826296
          BQ225468.1 - 5, 6 529
          CR627249.1 - 4, 5 7299

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0