Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142627.2 | NP_001136099.1 | 5, 6 | 498 | ||
NM_001142628.1 | NP_001136100.1 | 4, 5 | 432 | |||
NM_018144.3 | NP_060614.2 | 5, 6 | 498 | |||
NR_024576.2 | 5, 6 | 498 | ||||
NR_024577.2 | 6, 7 | 631 | ||||
GenBank mRNA | AF346603.1 | - | 5, 6 | 439 | ||
AK022640.1 | - | 5, 6 | 433 | |||
AK057532.1 | - | 6, 7 | 572 | |||
AK292295.1 | - | 5, 6 | 448 | |||
AK301841.1 | - | 4, 5 | 385 | |||
BC026179.1 | - | 5, 6 | 448 | 4823130 | ||
BP212860.1 | - | 5, 6 | 498 |